missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MPPED2 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-56450
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
MPPED2 Polyclonal specifically detects MPPED2 in Human samples. It is validated for Immunocytochemistry/Immunofluorescence.
Spécification
| MPPED2 | |
| Polyclonal | |
| Immunocytochemistry/Immunofluorescence 1-4 ug/ml | |
| 239FBchromosome 11 open reading frame 8, C11orf8dJ1024C24.1, D11S302E, dJ873F21.1, EC 3.1, FAM1B, Fetal brain protein 239, metallophosphoesterase domain containing 2, Metallophosphoesterase domain-containing protein 2, metallophosphoesterase MPPED2 | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| Immunocytochemistry, Immunofluorescence | |
| Unconjugated | |
| PBS, pH 7.2, containing 40% glycerol with 0.02% Sodium Azide | |
| MPPED2 | |
| This antibody was developed against a recombinant protein corresponding to the following amino acid sequence:MAHGIPSQGKVTITVDEYSS | |
| 100 μL | |
| Neuroscience | |
| 744 | |
| Human | |
| IgG |
Correction du contenu d'un produit
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit
For Research Use Only
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu