missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MPPED1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 13677-1-AP-150UL
This item is not returnable.
View return policy
Description
MPPED1 may have metallophosphoesterase activity (in vitro).Specifications
| MPPED1 | |
| Polyclonal | |
| Unconjugated | |
| MPPED1 | |
| 239AB, Adult brain protein 239, C22orf1, FAM1A, MPPED1 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 223726, 362971, 758 | |
| -20°C | |
| Liquid |
| Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
| 0.2 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| O15442, Q91ZG2 | |
| Mpped1 | |
| MPPED1 Fusion Protein Ag4672 | |
| 150 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction