missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ MPPE1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA552965
This item is not returnable.
View return policy
Description
Immunogen sequence: PEVKTTASDG EQTTREPVLK AMFLADTHLL GEFLGHWLDK LRREWQMERA FQTALWLLQP EVVFILGDIF DEGKWSTPEA WADDVERFQK MFRHPSHVQL KVVAGNHDIG FHYEMN Highest antigen sequence identity to the following orthologs: Mouse - 84%, Rat - 82%.
Metallophosphoesterase required for transport of GPI-anchor proteins from the endoplasmic reticulum to the Golgi. Acts in lipid remodeling steps of GPI-anchor maturation by mediating the removal of a side-chain ethanolamine-phosphate (EtNP) from the second Man (Man2) of the GPI intermediate, an essential step for efficient transport of GPI-anchor proteins. [UniProt].
Specifications
| MPPE1 | |
| Polyclonal | |
| Unconjugated | |
| MPPE1 | |
| A530095G11; metallo phosphoesterase; metallophosphoesterase 1; Mppe1; PGAP5; post-GPI attachment to proteins factor 5; PP579 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 65258 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 0.1 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q53F39 | |
| MPPE1 | |
| Recombinant protein corresponding to Human MPPE1. Recombinant protein control fragment (Product #RP-88831). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction