missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MPPE1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-05976-100ul
This item is not returnable.
View return policy
Description
MPPE1 Polyclonal antibody specifically detects MPPE1 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Specifications
| MPPE1 | |
| Polyclonal | |
| Western Blot 1:500-1:2000, Immunohistochemistry, Immunohistochemistry-Paraffin 1:50-1:200 | |
| EC 3.1, metallo phosphoesterase, metallophosphoesterase 1, PGAP5, Post-GPI attachment to proteins factor 5 | |
| Produced in rabbits immunized with E. coli-derived Human MPPE1 fragment. | |
| 100 μg | |
| Stem Cell Markers | |
| 65258 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| PBS, pH7.0 | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction