missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ MPP9 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA557970
This item is not returnable.
View return policy
Description
Immunogen sequence: YKSKDPKEFM EHIDVPKGQY VAPAVPAESL VDGVKNENFY IQTPEECHVS LKEDVSISPG EFEHNFLGEN KVSEVYSGKT NSNAITSWAQ KLKQNQPK Highest antigen sequence identity to the following orthologs: Mouse - 56%, Rat - 64%.
Progression of cells from interphase to mitosis involves alterations in cell structures and activities. The transition from G2 to M phase is induced by M phase-promoting factor (MPF). In M phase, many proteins are phosphorylated directly by MPF or indirectly by kinases activated by MPF. These M phase phosphoproteins (MPPs), also known as MPHOSPHs, permit disassembly of interphase structures and generation of M phase enzymatic activities and structures. MPP9 (M-phase phosphoprotein 9), also known as MPHOSPH9, is a 1,031 amino acid peripheral membrane protein of the Golgi apparatus that exists as two alternatively spliced isoforms. The gene encoding MPP9 maps to human chromosome 12, which encodes over 1,100 genes and comprises approximately 4.5% of the human genome. Chromosome 12 is associated with a variety of diseases and afflictions, including hypochondrogenesis, achondrogenesis, Kniest dysplasia, Noonan syndrome and trisomy 12p.
Specifications
| MPP9 | |
| Polyclonal | |
| Unconjugated | |
| MPHOSPH9 | |
| 4930548D04Rik; 9630025B04Rik; AW413446; AW547060; B930097C17Rik; C87456; M-phase phosphoprotein 9; MPHOSPH9; MPP9; MPP-9 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 10198 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 0.5 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q99550 | |
| MPHOSPH9 | |
| Recombinant protein corresponding to Human MPP9. Recombinant protein control fragment (Product #RP-97636). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction