missing translation for 'onlineSavingsMsg'
Learn More

MPP3 Antibody (OTI8F1), mFluor Violet 450 SE, Novus Biologicals™

Product Code. 18405793 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18405793 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18405793 Supplier Novus Biologicals Supplier No. NBP272771MFV450

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

MPP3 Monoclonal antibody specifically detects MPP3 in Human, Mouse, Rat samples. It is validated for Flow Cytometry
TRUSTED_SUSTAINABILITY

Specifications

Antigen MPP3
Applications Flow Cytometry
Classification Monoclonal
Clone OTI8F1
Conjugate mFluor Violet 450 SE
Dilution Flow Cytometry
Formulation 50mM Sodium Borate
Gene Alias Discs large homolog 3, discs, large homolog 3, DLG3MAGUK p55 subfamily member 3, membrane protein palmitoylated 3, membrane protein, palmitoylated 3 (MAGUK p55 subfamily member 3), Protein MPP3
Host Species Mouse
Immunogen Full length human recombinant protein of human MPP3 (NP_001923) produced in HEK293T cell.
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4356
Target Species Human, Mouse, Rat
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.