missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
MPP3 Polyclonal antibody specifically detects MPP3 in Human,Mouse,Rat samples. It is validated for ELISA,Immunohistochemistry
Specifications
Specifications
| Antigen | MPP3 |
| Applications | ELISA, Immunohistochemistry |
| Classification | Polyclonal |
| Conjugate | mFluor Violet 500 SE |
| Formulation | 50mM Sodium Borate |
| Gene Alias | Discs large homolog 3, discs, large homolog 3, DLG3MAGUK p55 subfamily member 3, membrane protein palmitoylated 3, membrane protein, palmitoylated 3 (MAGUK p55 subfamily member 3), Protein MPP3 |
| Host Species | Rabbit |
| Immunogen | membrane protein, palmitoylated 3(MAGUK p55 subfamily member 3). (Uniprot# Q13368) |
| Purification Method | Immunogen affinity purified |
| Quantity | 0.1 mL |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?