missing translation for 'onlineSavingsMsg'
Learn More

MPP3 Antibody (4D7), Novus Biologicals™

Product Code. 18383878 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mg
Unit Size:
0.01mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18383878 0.1 mg 0.01mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18383878 Supplier Novus Biologicals Supplier No. H00004356M01

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

MPP3 Monoclonal antibody specifically detects MPP3 in Human samples. It is validated for Western Blot, ELISA, ELISA, KnockDown
TRUSTED_SUSTAINABILITY

Specifications

Antigen MPP3
Applications Western Blot, ELISA, Sandwich ELISA, KnockDown
Classification Monoclonal
Clone 4D7
Conjugate Unconjugated
Dilution Western Blot 1:500, ELISA, Sandwich ELISA, Knockdown Validated
Formulation In 1x PBS, pH 7.4
Gene Accession No. AAH56865
Gene Alias Discs large homolog 3, discs, large homolog 3, DLG3MAGUK p55 subfamily member 3, membrane protein palmitoylated 3, membrane protein, palmitoylated 3 (MAGUK p55 subfamily member 3), Protein MPP3
Host Species Mouse
Immunogen MPP3 (AAH56865, 1 a.a. ~ 585 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. MPVLSEDSGLHETLALLTSQLRPDSNHKEEMGFLRDVFSEKSLSYLMKIHEKLRYYERQSPTPVLHSAVALAEDVMEELQAASVHSDERELLQLLSTPHLRAVLMVHDTVAQKNFDPVLPPLPDNIDEDFDEESVKIVRLVKNKEPLGATIRRDEHSGAVVVARIMRGGAADRSGLVHVGDELREVNGIAVLHKRPDEISQILAQSQGSITLKIIPATQEEDRLKESKVFMRALFHYNPREDRAIPCQEAGLPFQRRQVLEVVSQDDPTWWQAKRVGDTNLRAGLIPSKGFQERRLSYRRAAGTLPSPQSLRKPPYDQPCDKETCDCEGYLKGHYVAGLRRSFRLGCRERLGGSQEGKMSSGAESPELLTYEEVARYQHQPGERPRLVVLIGSLGARLHELKQKVVAENPQHFGVAVPHTTRPRKSHEKEGVEYHFVSKQAFEADLHHNKFLEHGEYKENLYGTSLEAIQAVMAKNKVCLVDVEPEALKQLRTSEFKPYIIFVKPAIQEKRKTPPMSPACEDTAAPFDEQQQEMAASAAFIDRHYGHLVDAVLVKEDLQGAYSQLKVVLEKLSKDTHWVPVSWVR
Purification Method IgG purified
Quantity 0.1 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4356
Target Species Human
Content And Storage Aliquot and store at -20°C or -80°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.