missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ MPP2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA521526
This item is not returnable.
View return policy
Description
Recommended positive controls: A549, HeLa, HCT116, mouse brain. Predicted reactivity: Mouse (98%), Rat (98%), Xenopus laevis (82%), Bovine (98%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
The MAGUK (membrane-associated guanylate kinase homologs) family of proteins contain multiple protein-binding domains and are involved in cell junction organization, tumor suppression and signaling. The MAGUK family is divided into four subfamilies: DLG-like, ZO1-like, p55-like and LIN2-like. MPP2 (membrane protein, palmitoylated 2), also known as MAGUK p55 subfamily member 2, discs large homolog 2 or DLG2, is a 576 amino acid protein belonging to the MAGUK family that exists as three alternatively spliced isoforms. MPP2 contains one guanylate kinase-like domain, a PDZ (DHR) domain, two L27 domains and a single SH3 domain. The gene encoding MPP2 maps to the same segment of human chromosome 17 as MPP3, with whom MMP2 likely shares similar function and common structural organization.
Specifications
| MPP2 | |
| Polyclonal | |
| Unconjugated | |
| Mpp2 | |
| D11Bwg0652e; discs large homolog 2; discs large, homolog 2; Dlg2; Dlgh2; MAGUK p55 subfamily member 2; membrane palmitoylated protein 2; membrane protein, palmitoylated 2 (MAGUK p55 subfamily member 2); Mpp2; p55 subfamily member of MAGUK family; palmitoylated membrane protein 2; Pals4; Protein MPP2; rCG_34691 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 4355, 50997 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot | |
| 0.61 mg/mL | |
| 0.1M tris glycine with 10% glycerol and 0.01% thimerosal; pH 7 | |
| Q14168, Q9WV34 | |
| Mpp2 | |
| Recombinant protein encompassing a sequence within the center region of human MPP2. The exact sequence is proprietary. | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction