missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ MPP1 Recombinant Rabbit Monoclonal Antibody (23GB4770), Invitrogen™
GREENER_CHOICE

Rabbit Recombinant Monoclonal Antibody

Brand:  Invitrogen™ MA552734

 View more versions of this product

Product Code. 30089103

  • £387.00 / 100µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

This gene encodes the prototype of the membrane-associated guanylate kinase (MAGUK) family proteins. MAGUKs interact with the cytoskeleton and regulate cell proliferation, signaling pathways, and intercellular junctions. The encoded protein is an extensively palmitoylated membrane phosphoprotein containing a PDZ domain, a Src homology 3 (SH3) motif, and a guanylate kinase domain. This gene product interacts with various cytoskeletal proteins and cell junctional proteins in different tissue and cell types, and may be involved in the regulation of cell shape, hair cell development, neural patterning of the retina, and apico-basal polarity and tumor suppression pathways in non-erythroid cells. Multiple transcript variants encoding different isoforms have been found for this gene.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

MPP1
Recombinant Monoclonal
0.25 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
P70290, Q00013
MPP1
A synthesized peptide derived from human MPP1 (305-350AA).
100 μL
Primary
Human, Mouse
Antibody
IgG
Western Blot, Immunocytochemistry
23GB4770
Unconjugated
MPP1
55 kDa erythrocyte membrane protein; 55kDa; AAG12; aging-associated gene 12; C130070C03Rik; DXS552E; EMP55; erythrocyte membrane protein p55; LOC652956; membrane palmitoylated protein 1; membrane protein, palmitoylated; membrane protein, palmitoylated 1; membrane protein, palmitoylated 1, 55kDa; migration-related gene 1; MPP1; MRG1; p55; p55 protein; palmitoylated erythrocyte membrane protein; PEMP
Rabbit
Affinity chromatography
RUO
17524, 4354
-20°C
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.