missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MPP1 Antibody (S03-1F4), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-19893-50ul
This item is not returnable.
View return policy
Description
MPP1 Monoclonal antibody specifically detects MPP1 in Human, Mouse samples. It is validated for Western Blot
Specifications
| MPP1 | |
| Monoclonal | |
| 0.3 mg/mL | |
| 50mM Tris-Glycine(pH 7.4), 0.15M NaCl, 40% Glycerol, 0.05% BSA | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse | |
| Purified |
| Western Blot | |
| S03-1F4 | |
| Unconjugated | |
| 55 kDa erythrocyte membrane protein, AAG12, EC 2.7.11, Membrane protein, palmitoylated 1, membrane protein, palmitoylated 1, 55kDa, MRG1, palmitoylated erythrocyte membrane protein, PEMPpalmitoylated 1 (55kD) | |
| A synthetic peptide of human MPP1 (Uniprot # Q00013) | |
| 50 μg | |
| Signal Transduction | |
| 4354 | |
| Store at 4°C short term. Store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction