missing translation for 'onlineSavingsMsg'
Learn More

mPlum Antibody, HRP, Novus Biologicals™

Product Code. 18374673 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18374673 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18374673 Supplier Novus Biologicals Supplier No. NBP311833H

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

mPlum Polyclonal antibody specifically detects mPlum in Nonspecies specific samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen mPlum
Applications Western Blot
Classification Polyclonal
Conjugate HRP
Formulation PBS
Gene Alias Fluorescent protein plum, mPlum Fluorescent Protein, Red Fluorescent Protein, RFP
Host Species Rabbit
Immunogen mPlum from Discosoma sp.
Purification Method Affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Target Species Non-species specific
Content And Storage Store at 4°C in the dark.
Form Purified
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.