Learn More
Invitrogen™ MPI Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA521510
Description
Recommended positive controls: 293T, A431, H1299, HeLaS3, HepG2, Molt-4, Raji, PC-12, Rat2. Predicted reactivity: Mouse (84%), Rat (86%), Pig (92%), Chimpanzee (99%), Bovine (92%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
Phosphomannose isomerase catalyzes the interconversion of fructose-6-phosphate and mannose-6-phosphate and plays a critical role in maintaining the supply of D-mannose derivatives, which are required for most glycosylation reactions. Mutations in the MPI gene were found in patients with carbohydrate-deficient glycoprotein syndrome, type Ib.
Specifications
| MPI | |
| Polyclonal | |
| Unconjugated | |
| MPI | |
| 1110002E17Rik; AI315153; CDG1B; mannose phosphate isomerase; mannose phosphate isomerase (mapped); mannose phosphate isomerase 1; mannose-6-phosphate isomerase; MPI; Mpi1; Mpi-1; Phosphohexomutase; phosphomannose isomerase; phosphomannose isomerase 1; PMI; PMI1 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 300741, 4351 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 1 mg/mL | |
| 0.1M tris glycine with 10% glycerol and 0.01% thimerosal; pH 7 | |
| P34949, Q68FX1 | |
| MPI | |
| Recombinant protein encompassing a sequence within the center region of human MPI. The exact sequence is proprietary. | |
| 100 μL | |
| Primary | |
| Human, Rat | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.