missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ MPI Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA582287
This item is not returnable.
View return policy
Description
Immunogen sequence: GSNSEVARLL ASSDPLAQIA EDKPYAELWM GTHPRGDAKI LDNRISQKTL SQWIAENQDS LGSKVKDTFN GNLPFLFKVL SVETPLSIQA HPNKELAEKL HLQAPQHYPD ANHKPEMAIA LTPFQGLCG Highest antigen sequence indentity to the following orthologs: Rat - 91%, Mouse - 91%.
Phosphomannose isomerase catalyzes the interconversion of fructose-6-phosphate and mannose-6-phosphate and plays a critical role in maintaining the supply of D-mannose derivatives, which are required for most glycosylation reactions. Mutations in the MPI gene were found in patients with carbohydrate-deficient glycoprotein syndrome, type Ib.
Specifications
| MPI | |
| Polyclonal | |
| Unconjugated | |
| MPI | |
| 1110002E17Rik; AI315153; CDG1B; mannose phosphate isomerase; mannose phosphate isomerase (mapped); mannose phosphate isomerase 1; mannose-6-phosphate isomerase; MPI; Mpi1; Mpi-1; Phosphohexomutase; phosphomannose isomerase; phosphomannose isomerase 1; PMI; PMI1 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 4351 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.1 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| P34949 | |
| MPI | |
| Recombinant protein corresponding to Human MPI. Recombinant protein control fragment (Product #RP-89702). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction