missing translation for 'onlineSavingsMsg'
Learn More

MPHOSPH6 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Product Code. 16877243 Shop All Proteintech Products
Change view
Click to view available options
Quantity:
20 μL
150 μL
Unit Size:
150µL
20µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16877243 20 μL 20µL
16867243 150 μL 150µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16877243 Supplier Proteintech Supplier No. 106951AP20UL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

RNA-binding protein that associates with the RNA exosome complex. Involved in the 3′-processing of the 7S pre-RNA to the mature 5. 8S rRNA and play a role in recruiting the RNA exosome complex to pre-rRNA; this function may include C1D.

Specifications

Antigen MPHOSPH6
Applications Immunoprecipitation, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.33 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Gene MPHOSPH6
Gene Accession No. Q99547, Q9D1Q1
Gene Alias M phase phosphoprotein 6, MPHOSPH6, MPP, MPP 6, MPP6
Gene Symbols Mphosph6
Host Species Rabbit
Immunogen MPHOSPH6 Fusion Protein Ag1080
Purification Method Antigen Affinity Chromatography
Quantity 20 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10200, 68533, 686999
Target Species Human, Rat, Mouse
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.