missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ MPHOSPH6 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5109972
This item is not returnable.
View return policy
Description
Immunogen sequence: MAAERKTRLS KNLLRMKFMQ RGLDSETKKQ LEEEEKKIIS EEHWYLDLPE LKEKESFIIE EQSFLLCEDL LYGRMSFRGF NPEVEKLMLQ MNAKHKAEEV EDETVELDVS DEEMARRYET LVGTIGKKFA RKRDHANYEE DENGDITPIK AKKMFLKPQD.
MPHOSPH6 (M Phase Phosphoprotein 6) is a Protein Coding gene. Among its related pathways areGene ExpressionandDeadenylation dependent mRNA decay. Gene Ontology (GO) annotations related to this gene includeRNA binding.
Specifications
| MPHOSPH6 | |
| Polyclonal | |
| Unconjugated | |
| MPHOSPH6 | |
| 1110001M01Rik; AA536809; C86426; M phase phosphoprotein 6; M-phase phosphoprotein 6; Mphosph6; MPP; MPP6; MPP-6 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 10200, 68533, 686999 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Western Blot | |
| 2.10 mg/mL | |
| PBS with 50% glycerol and 0.01% thimerosal; pH 7.3 | |
| Q99547, Q9D1Q1 | |
| MPHOSPH6 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-160 of human MPHOSPH6 (NP_005783.2). | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction