missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MPHOSPH6 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-35598-100ul
This item is not returnable.
View return policy
Description
MPHOSPH6 Polyclonal antibody specifically detects MPHOSPH6 in Human,Mouse,Rat samples. It is validated for ELISA,Western Blot
Specifications
| MPHOSPH6 | |
| Polyclonal | |
| Western Blot 1:500 - 1:2000, ELISA | |
| M-phase phosphoprotein 6, MPP-6, MPP6MPP | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-160 of human MPHOSPH6 (NP_005783.2).,, Sequence:, MAAERKTRLSKNLLRMKFMQRGLDSETKKQLEEEEKKIISEEHWYLDLPELKEKESFIIEEQSFLLCEDLLYGRMSFRGFNPEVEKLMLQMNAKHKAEEVEDETVELDVSDEEMARRYETLVGTIGKKFARKRDHANYEEDENGDITPIKAKKMFLKPQD | |
| 100 μL | |
| Cell Cycle and Replication | |
| 10200 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| ELISA, Western Blot | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction