missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
MPHOSPH1 Polyclonal specifically detects MPHOSPH1 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
Specifications
| Antigen | MPHOSPH1 |
| Applications | Immunohistochemistry, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | DyLight 350 |
| Dilution | Immunohistochemistry, Immunohistochemistry-Paraffin |
| Formulation | 50mM Sodium Borate |
| Gene Alias | Cancer/testis antigen 90, CT90MPP-1, DKFZp434B0435, EC 6.3.5.5, kinesin family member 20B, Kinesin-related motor interacting with PIN1, KRMP1kinesin-like protein KIF20B, M-phase phosphoprotein 1mitotic kinesin-like protein, MPHOSPH1EC 2.1.1, MPP1DKFZp434P0810 |
| Host Species | Rabbit |
| Immunogen | Produced in rabbits immunized with E. coli-derived Human MPHOSPH1 fragment. |
| Purification Method | Antigen and protein A Affinity-purified |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?