missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Spectrum Labs™ mPES MiniKros™ Filter Modules
Spectrum Labs™ mPES MiniKros™ Filter Modules hollow fiber filters designed for tangential flow filtration.
£1575.00 - £1580.00
Specifications
Material | Modified PES |
---|---|
Sterility | Non-sterile |
Includes | Inlet/Outlet Ports: 1.5 in. TC, Side Port: 0.75 in. TC |
Type | Hollow Fiber Filtration Module |
Length (Metric) | Total Length: 70cm, Effective Length: 65cm |
Product Code | Brand | Description | Diameter (Metric) Inside | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Description | Diameter (Metric) Inside | Price | Quantity & Availability | ||||
15800790
|
Spectrum Labs™
N06-E030-05-N |
Surface area: 0.85m2, MWCO: 30 kDa | Fiber ID: 0.5mm |
£1575.00
Each |
Estimated Shipment: 05-07-2024
Log in to see stock. |
||||
15810790
|
Spectrum Labs™
N06-E65U-07-N |
Surface area: 0.61m2, Pore Size: 0.65 μm | Fiber ID: 0.75mm |
£1580.00
Each |
Estimated Shipment: 05-07-2024
Log in to see stock. |
||||
Description
- Designed for tangential flow filtration of small volumes ranging from 5L to 50L
- Hollow fiber filters are suitable for pilot scale applications
- Come with 1-1/2 in. TC x 3/4 in. TC connections and are available in different lengths
Specifications
Modified PES | |
Inlet/Outlet Ports: 1.5 in. TC, Side Port: 0.75 in. TC | |
Total Length: 70cm, Effective Length: 65cm |
Non-sterile | |
Hollow Fiber Filtration Module | |
Housing OD: 33.1 mm |