missing translation for 'onlineSavingsMsg'
Learn More

MPEP Hydrochloride, MedChemExpress

Product Code. 16472247
Encompass_Preferred
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
This item is not returnable. View return policy

Product Code. 16472247

Brand: Medchem Express HY14609

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress MPEP Hydrochloride is a potent, selective, noncompetitive, orally active and systemically active mGlu5 receptor antagonist, with an IC50 of 36 nM for completely inhibiting quisqualate-stimulated phosphoinositide (PI) hydrolysis. MPEP Hydrochloride has anxiolytic-or antidepressant-like effects.

MPEP Hydrochloride is a potent, selective, noncompetitive, orally active and systemically active mGlu5 receptor antagonist, with an IC50 of 36 nM for completely inhibiting quisqualate-stimulated phosphoinositide (PI) hydrolysis. MPEP Hydrochloride has anxiolytic-or antidepressant-like effects.

Specifications

Chemical Name or Material MPEP Hydrochloride
CAS 219911-35-0
Color White
Purity Grade Notes Research
Molecular Formula C14H12ClN
Quantity 50 mg
Solubility Information H2O : ≥ 50 mg/mL (217.68 mM)
SMILES CC1=NC(C#CC2=CC=CC=C2)=CC=C1.[H]Cl
Molecular Weight (g/mol) 229.7
Formula Weight 229.7
Percent Purity 99.69%
Grade Research
Physical Form Solid
For Use With (Application) Neuroscience-Neurodegeneration
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.