missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MPEP Hydrochloride, MedChemExpress
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
Description
MPEP Hydrochloride is a potent, selective, noncompetitive, orally active and systemically active mGlu5 receptor antagonist, with an IC50 of 36 nM for completely inhibiting quisqualate-stimulated phosphoinositide (PI) hydrolysis. MPEP Hydrochloride has anxiolytic-or antidepressant-like effects.
Specifications
Specifications
| Chemical Name or Material | MPEP Hydrochloride |
| CAS | 219911-35-0 |
| Color | White |
| Purity Grade Notes | Research |
| Molecular Formula | C14H12ClN |
| Quantity | 10 mg |
| Solubility Information | H2O : ≥ 50 mg/mL (217.68 mM) |
| SMILES | CC1=NC(C#CC2=CC=CC=C2)=CC=C1.[H]Cl |
| Molecular Weight (g/mol) | 229.7 |
| Formula Weight | 229.7 |
| Show More |
Research purposes only
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction