missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ MPEP hydrochloride
GREENER_CHOICE

Product Code. 18779896
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
This item is not returnable. View return policy

Product Code. 18779896

Brand: Tocris Bioscience™ 1212/50

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Potent mGlu5 antagonist; also positive allosteric modulator of mGlu4 receptors

MPEP hydrochloride is a potent and highly selective non-competitive antagonist at the mGlu5 receptor subtype (IC50 = 36 nM) and a positive allosteric modulator at mGlu4 receptors. Centrally active following systemic administration in vivo. Reverses mechanical hyperalgesia in the inflamed rat hind paw.
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 2-Methyl-6-(phenylethynyl)pyridine hydrochloride
CAS 219911-35-0
Assay Percent Range >99%
Quantity 50 mg
Solubility Soluble to 100mM in DMSO
Target Group I mGluR Antagonists
Molecular Formula C14H11N.HCl
Formula Weight Observed MW: 229.71
Purity 99%

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.