missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ MPEG1 Polyclonal Antibody, Invitrogen™

Rabbit Polyclonal Antibody

Brand:  Invitrogen™ PA5121769

 View more versions of this product

Product Code. 17773164

  • £133.00 / 20µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

The MPEG1 (Macrophage Expressed Gene 1) gene encodes a protein known as Perforin-2, which is part of the immune system and plays a crucial role in host defense mechanisms. Located on chromosome 11, MPEG1/Perforin-2 is primarily expressed in macrophages, dendritic cells, and other immune cells. The protein functions by forming pores in the membranes of pathogenic bacteria, leading to their lysis and destruction. This pore-forming capability is integral to the innate immune response, providing a rapid means to target and eliminate intracellular pathogens. MPEG1's activity contributes significantly to the antimicrobial arsenal of the innate immune system, and deficiencies in this protein can lead to increased susceptibility to infections. Ongoing research is focused on understanding how MPEG1/Perforin-2 can be leveraged or augmented to enhance immune responses and potential therapeutic applications in infectious disease and immunotherapy.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

MPEG1
Polyclonal
Unconjugated
Mpeg1
macrophage expressed 1; macrophage expressed gene 1; macrophage gene 1 protein; Macrophage-expressed gene 1 protein; MGC150002; MGC150003; MPEG1; MPG1; Mpg-1; MPS1; P-2; perforin 2; Perforin-2; Protein MPS1
Rabbit
Antigen affinity chromatography, Protein A
RUO
219972
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Immunohistochemistry (Paraffin), Western Blot
0.5 mg/mL
PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4
Q2M385
Mpeg1
Phe313-Ser580 (Accession Q2M385), with N-terminal His Tag.
20 μL
Primary
Human
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.