missing translation for 'onlineSavingsMsg'
Learn More

MPEG-2000-DSPE, TRC

Product Code. 17941245
Change view
Click to view available options
Quantity:
100 mg
250 mg
500 mg
Unit Size:
100mg
250mg
500mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17941245 250 mg 250mg
17935055 100 mg 100mg
17932841 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 17941245 Supplier Toronto Research Chemicals Supplier No. TRCM745845250MG
 Restricted Item
Estimated Shipment: 28-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 147867-65-0
Molecular Formula (C2H4O)nC43H84NO10P
Molecular Weight (g/mol) 872.1
Synonym α-[6-Hydroxy-6-oxido-1,12-dioxo-9-[(1-oxooctadecyl)oxy]-5,7,11-trioxa-2-aza-6-phosphanonacos-1-yl]-ω-methoxy-Poly(oxy-1,2-ethanediyl)
IUPAC Name ethyl (E,4S)-2-methyl-4-[(2-methylpropan-2-yl)oxycarbonylamino]-5-(4-phenylphenyl)pent-2-enoate
SMILES CC(C)(C)OC(N[C@H](/C=C(C)/C(OCC)=O)CC(C=C1)=CC=C1C2=CC=CC=C2)=O

Specifications

Quantity 250 mg
InChI Formula InChI=1S/C25H31NO4/c1-6-29-23(27)18(2)16-22(26-24(28)30-25(3,4)5)17-19-12-14-21(15-13-19)20-10-8-7-9-11-20/h7-16,22H,6,17H2,1-5H3,(H,26,28)/b18-16+/t22-/m1/s1
Formula Weight 409.2253
Chemical Name or Material MPEG-2000-DSPE
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.