missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ MPD 2

Product Code. 30279879
Click to view available options
:
5 mg
This item is not returnable. View return policy

Product Code. 30279879

Brand: Tocris Bioscience™ 8802/5

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

SARS-CoV-2 Mpro Degrader (PROTAC™)

MPD 2 is a SARS-CoV-2 Mpro Degrader (PROTAC™) (DC50 = 296 nM in 293T cells). Mpro degradation is time-dependent and CRBN-mediated. Antiviral in A549-ACE2 cells infected with several SARS-CoV-2 strains (EC50 = 492 nM at delta variant) and against a nirmatrelvir-resistant mutant of SARS-CoV-2, NSP5 E166A (EC50 = 460 nM).

PROTAC™ is a registered trademark of Arvinas Operations, Inc., and is used under license.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 3-((6-((2-(2,6-Dioxopiperidin-3-yl)-1,3-dioxoisoindolin-4-yl)oxy)hexyl)oxy)benzyl ((2S,3R)-3-(tert-butoxy)-1-(((S)-3-cyclohexyl-1-oxo-1-(((S)-1-oxo-3-((S)-2-oxopyrrolidin-3-yl)propan-2-yl)amino)propan-2-yl)amino)-1-oxobutan-2-yl)carbamate
Quantity 5 mg
Target Active Degraders
Molecular Formula C51H68N6O13
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.