Learn More
CPC (Colder) MPCTM Quick-Disconnect Fittings, Plug
Product Code. p-200105151
Click to view available options
Color:
Amber tint
Purple Tint
Unit Size:
Pack of 25
Product Code. 30053149
Brand: CPC (Colder) 5011274
Only null left
Add to basket
Add to basket
Connect to a female coupling body to stop the flow
Ideal for general-purpose, laboratory, and single-use applications to connect tubing with the rest of your system. Free technical application support available to determine which connector type is needed for your application
Specifications
Certifications/Compliance ADCF;FDA;Non-DEHP;Non-Phthalate;RoHS;USP
Min. Operating Temperature (Metric) -40°F
Outlet Port Plug
Shape Plug
Material (Seal) Platinum-cured Silicone
Color Purple Tint
Warning: This product is not approved or intended for, and should not be used for, medical, clinical, surgical or other patient-oriented applications.