Learn More
CPC (Colder) MPC/MPX Quick-Disconnect Fittings, Back-to-Back Body Adapter
Product Code. 30053892
Click to view available options
Color:
Amber tint
Purple Tint
Outlet Port:
MPC Body x MPC Body
MPC Body x MPX Body
MPX Body x MPX Body
Unit Size:
Each
Product Code. 30053892
Brand: CPC (Colder) 5011311
Only null left
Add to basket
Add to basket
Connect single-use systems that feature identical coupling connections at the end of their tubing
Ideal for general-purpose, laboratory, and single-use applications to connect tubing with the rest of your system. Free technical application support available to determine which connector type is needed for your application. Connect to a male coupling insert for easy media transfer. Operate ergonomic thumb latch even with gloved hands. Combine MPC and MPX couplings for reducing options
Specifications
Certifications/Compliance ADCF;FDA;Non-DEHP;Non-Phthalate;RoHS;USP
Min. Operating Temperature (Metric) -40°F
Outlet Port MPX Body x MPX Body
Sterilization Method 121°C
Shape Straight
Color Purple Tint