missing translation for 'onlineSavingsMsg'
Learn More

MP7, MedChemExpress

Product Code. 16476068
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16476068 10 mg 10mg
16466466 100 mg 100mg
16462207 50 mg 50mg
16438677 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 16476068 Supplier Medchem Express Supplier No. HY14440
 Restricted Item
Estimated Shipment: 30-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress MP7 (PDK1 inhibitor) is a phosphoinositide-dependent kinase-1 (PDK1) inhibitor.

MP7 (PDK1 inhibitor) is a phosphoinositide-dependent kinase-1 (PDK1) inhibitor.

Specifications

Chemical Name or Material MP7
CAS 1001409-50-2
Color Yellow
Purity Grade Notes Research
Molecular Formula C28H22F2N4O4
Quantity 10 mg
Synonym PDK1 inhibitor
Solubility Information DMSO : ≥ 100 mg/mL (193.61 mM)
SMILES O=C(C1=CC=CN(CC2=CC=C(F)C(F)=C2)C1=O)N[C@H](C3=CC=CC=C3)COC4=CC=C(N5)C(NC5=O)=C4
Molecular Weight (g/mol) 516.5
Formula Weight 516.5
Percent Purity 99.83%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.