missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ MP68 Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA563868
This item is not returnable.
View return policy
Description
Immunogen sequence: MKLYGTRGSD ISHFQCQMLQ SIIK Highest antigen sequence identity to the following orthologs: Mouse - 46%, Rat - 46%.
MP68, also known as PLPM or C14orf2 (chromosome 14 open reading frame 2), is a 58 amino acid mitochondrial protein that belongs to the small mitochondrial proteolipid family. The gene encoding MP68 maps to human chromosome 14, which houses over 700 genes and comprises nearly 3.5% of the human genome. Chromosome 14 encodes the presinilin 1 (PSEN1) gene, which is one of the three key genes associated with the development of Alzheimer's disease (AD). The SERPINA1 gene is also located on chromosome 14 and, when defective, leads to the genetic disorder alpha1-antitrypsin deficiency, which is characterized by severe lung complications and liver dysfunction.
Specifications
| MP68 | |
| Polyclonal | |
| Unconjugated | |
| Atp5mpl | |
| 2010107E04Rik; 6.8 kDa mitochondrial proteolipid; 6.8 kDa mitochondrial proteolipid protein; 9430003J03Rik; ATP synthase membrane subunit 6.8PL; ATP synthase subunit ATP5MPL, mitochondrial; Atp5mpl; AU043134; AV124504; C14orf2; chromosome 14 open reading frame 2; MLQ; Mp68; PLPM; PRO1574; RIKEN cDNA 2010107E04 gene | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 9556 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 0.1 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| P56378 | |
| Atp5mpl | |
| Recombinant protein corresponding to Human MP68. Recombinant protein control fragment (Product #RP-100879). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction