missing translation for 'onlineSavingsMsg'
Learn More

MP1/MAP2K1IP1 Antibody, Alexa Fluor™ 405, Novus Biologicals™

Product Code. 18424997 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.10mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18424997 0.1 mL 0.10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18424997 Supplier Novus Biologicals Supplier No. NBP281886AF405

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

MP1/MAP2K1IP1 Polyclonal antibody specifically detects MP1/MAP2K1IP1 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY

Specifications

Antigen MP1/MAP2K1IP1
Applications Western Blot, ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Alexa Fluor 405
Formulation 50mM Sodium Borate
Gene Alias Late endosomal/lysosomal adaptor and MAPK and MTOR activator 3, late endosomal/lysosomal adaptor, MAPK and MTOR activator 3, MAP2K1IP1, MAPBP, MAPK scaffold protein 1, MAPKSP1, MEK binding partner 1, MEK partner 1, MEK-binding partner 1, mitogen-activated protein kinase kinase 1 interacting protein 1, Mitogen-activated protein kinase kinase 1-interacting protein 1, Mitogen-activated protein kinase scaffold protein 1, Mp1, PRO0633, Ragulator3
Host Species Rabbit
Immunogen MAP2K1IP1 antibody was raised against an 15 amino acid peptide near the center of human MAP2K1IP1. The immunogen is located within amino acids 40 - 90 of MAP2K1IP1. Amino Acid Squence: ATDQGSKLGLSKNKS
Purification Method Peptide affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 8649
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.