missing translation for 'onlineSavingsMsg'
Learn More

MP EcoChrom Aluminum Oxide, Acid Technical A , MP Biomedicals, LLC

Product Code. 15576226 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
100 kg
Unit Size:
100kg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15576226 100 kg 100kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15576226 Supplier MP Biomedicals Supplier No. 0207896

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MP EcoChrom Aluminum Oxide, Acid Technical A is a white to light pink free flowing powder that is ideal for quality control.

  • Ideal for the isolation of alkaloids, glycosides, active substances from plants, vitamins, hormones, and antibiotics
  • Dehydration of organic solvents
  • Purification of enzymes
  • Removal of terpenes, peroxides, and pyrogens from injectable solutions and infusions
TRUSTED_SUSTAINABILITY

Specifications

Boiling Point ∼2980°C
Melting Point ∼2072°C
Color Pink to White
Quantity 100 kg
Density 3.94 g/cm³at 20 °C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.