missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MP Biomedicals™ MP EcoChrom Alumina N, Neutral pH (for HPLC)
Shop All MP Biomedicals Products
Click to view available options
:
Particle Size: 10 to 18 μm
Description
- Usage Statement: Unless specified otherwise, MP Biomedical's products are for research or further manufacturing use only, not for direct human use.
Specifications
Specifications
| Particle Size | 10 to 18 μm |
| Technique | Liquid Chromatography |
| pH | Neutral |
| Type | Alumina N |
| Material | Alumina |
| For Use With (Application) | HPLC |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction