missing translation for 'onlineSavingsMsg'
Learn More

MP Biomedicals™ Tryptic Soy Agar

Product Code. 11470245 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
500 g
Unit Size:
500g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11470245 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11470245 Supplier MP Biomedicals™ Supplier No. 091010617

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Used for the cultivation of fastidious and non-fastidious microorganisms.

Tryptic Soy Agar is a medium very rich in nutrients for 'general use' in microbiological laboratories.

Containing two peptones obtained by enzymatic hydrolysis of casein and soy protein, this medium supports the growth of a great variety of microorganisms, including fastidious aerobes and anaerobes.

If desired, antibiotics can easily be incorporated as well as other supplements or inhibitory agents.

A short list of microorganisms that grow on this medium are the following: Streptococcus, Neisseria, Brucella, Corynebacterium, Listeria, Pasteurella, Vibrio, Haemophilus vaginals, Candida, etc.

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage 15 to 30°C
Form Powder
Media Type Tryptic Soy Agar
Product Type Tryptic Soy Agar
Formulation Casein Peptone 15 g/L, Sodium Chloride 5 g/L, Soy Peptone 5 g/L, Agar 15 g/L
For Use With (Application) Isolation and cultivation of fastidious organisms
Quantity 500 g

Unless specified otherwise, MP Biomedical's products are for research or further manufacturing use only, not for direct human use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.