missing translation for 'onlineSavingsMsg'
Learn More

MP Biomedicals™ TallPrep™ Tubes, 4.5 mL, Empty

Product Code. 11451565 Shop All MP Biomedicals Products
Click to view available options
Quantity:
100 Pk.
Unit Size:
Pack of 100
This item is not returnable. View return policy

Product Code. 11451565

Brand: MP Biomedicals™ 116970000

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Empty TallPrep™ Tubes for use in Nucleic Acid Isolation and Purification.

  • 4.5 mL TallPrep™ Tubes (Empty)
  • Caps included
  • Store at room temperature
TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Store at room temperature.
Sample Type All
Includes Caps
Product Type Empty Tube
Quantity 100 Pk.
Volume (Metric) 4.5 mL

Unless specified otherwise, for research or further manufacturing use only, not for direct human use.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.