missing translation for 'onlineSavingsMsg'
Learn More

MP Biomedicals™ TallFlex™ Adapter for 48 x 4.5 mL Tube Holder on FastPrep 96

Product Code. 15242820 Shop All MP Biomedicals Products
Click to view available options
For Use With (Equipment):
48 x 4.5 mL Tube Holder
Unit Size:
Pack of 48
This item is not returnable. View return policy

Product Code. 15242820

Brand: MP Biomedicals™ 116010580

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

TallFlex™ Adapter for 48 x 4.5 mL Tube Holder on FastPrep 96 is used for room temperature grinding.

  • Product Overview: Medium-scale adapter for the FastPrep-96™ that holds 48 x 4.5 mL Lysing Matrix Tubes. The ergonomic design of the adapter ensures an easy loading of Lysing Matrix tubes that remain securely sealed during the homogenization. It stands stable on the bench top and is commonly used as a tube rack. Additionally, frozen samples in Lysing Matrix tubes loaded in the adapter and stored at -20°C or -80°C are ready to be immediately processed with minimal hands-on manipulation, preventing degradation of cellular components by endogenous enzymes.
  • Application: TallFlex™ 48 x 4.5 mL Adapter for FastPrep-96™ is a large sample volume adapter is ideally suited for DNA and RNA isolation, enzyme isolation and protein production, natural products isolation, food preparation for quality analysis, biopharma manufacturing, and forensic applications. Additionally, frozen samples in Lysing Matrix tubes loaded in the adapter and stored at -20°C or -80°C are ready to be immediately processed with minimal hands-on manipulation, preventing degradation of cellular components by endogenous enzymes.
  • Capacity: 48 tubes
  • Sample Volume: Up to 20 g
TRUSTED_SUSTAINABILITY

Specifications

Product Type FastPrep-96 Adapters
Content And Storage Room Temperature
For Use With (Application) Extractions of nucleic acids, proteins, metabolites and other small molecules from samples
For Use With (Equipment) 48 x 4.5 mL Tube Holder

Unless specified otherwise, MP Biomedical's products are for research or further manufacturing use only, not for direct human use.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.