missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MP Biomedicals™ Streptavidin
Shop All MP Biomedicals Products
Click to view available options
Quantity:
5 mg
Unit Size:
5mg
Description
Has four binding sites for biotin, as does avidin. Both have been used extensively as probes in immunochemical systems, conjugated to antibodies, enzymes or fluorochromes.
- Format: Off-white clumpy powder
- Extinction Coefficient (E1%): E1% = 34 at 280nm (solvent: 0.1 M ammonium perchlorate, pH 8.0)(Lit.)
- Specific Activity: ∽10units/mg protein
Specifications
Specifications
| Molecular Weight (g/mol) | 55000 |
| Quantity | 5 mg |
| Source | Streptomyces avidinii |
| Content And Storage | 0°C |
| Form | Lyophilized |
| Product Type | Streptavidin from Streptomyces avidinii |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction