missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
- Known as Hopkins Mixture
- Available in complete formulations
- Available in dropout formulations
- Add 2g of SC to each liter of DOB or DOBA and autoclave at 121°C for 15 minutes for complete synthetic defined medium for S.cerevisiae
Add 2g of SC to each liter of DOB or DOBA and autoclave at 121°C for 15 minutes for complete synthetic defined medium for S.cerevisiae
Applications:
Yeast cell culture, Gene expression, Transcription regulation
Specifications
Specifications
| Certifications/Compliance | Molecular biology Certified™ |
| Content And Storage | 2°C to 8°C |
| Form | Powder |
| Product Type | Yeast Media |
| Formulation | Adenine 21mg/L, L-Alanine 86.5mg/L, L-Arginine HCl 85.6mg/L, L-Asparagine 85.6mg/L, L-Aspartic Acid 85.6mg/L, L-Cysteine HCl 85.6mg/L, Glutamine 85.6mg/L, L-Glutamic Acid 85.6mg/L, Glycine 85.6mg/L, L-Histidine HCl 85.6mg/L, Myo-Inositol 85.6mg/L, L-Isoleucine 85.6mg/L, L-Leucine 173.4mg/L, L-Lysine HCl 85.6mg/L, L-Methionine 85.6mg/L, Para-Aminobenzoic Acid 8.6mg/L, L-Phenylalanine 85.6mg/L, L-Proline 85.6mg/L, L-Serine 85.6mg/L, L-Threonine 85.6mg/L, L-Tryptophan 85.6mg/L, L-Tyrosine 85.6mg/L, Uracil 85.6mg/L, L-Valine 85.6mg/L |
| For Use With (Application) | For growth of Saccharomyces cerevisiae |
| Quantity | 30 g |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?