missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
- Kit is designed to isolate and purify RNA (75 bases to 10 kb) from native and denaturing agarose and polyacrylamide gels.
- Removal of unincorporated nucleotides, buffer salts, and other reagents from enzyme reactions.
- The purified RNA is ready for reverse transcription and PCR, labeling and microarray analysis, and other common applications.
Specifications
Specifications
| Sample Type | Enzymatic Reactions, Solutions, Agarose Gels, Polyacrylamide Gels |
| For Use With (Application) | To isolate, purify RNA (75 bases to 10 kb) from native, denaturing agarose/polyacrylamide gels |
| Product Type | Slurry plus SPIN Filter |
| Quantity | 200 Preps |
| Sample Source | Gels and Solutions |
Safety and Handling
Product Identifier
- RNaid Spin Kit
- Warning
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
- P101-If medical advice is needed, have product container or label at hand.
- P102-Keep out of reach of children.
- P103-Read label before use.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P321-Specific treatment (see on this label).
- P405-Store locked up.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
- MIXTURE LIST-Contain: diethyl pyrocarbonate
Unless specified otherwise, for research or further manufacturing use only, not for direct human use.
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?