missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MP Biomedicals™ Pyruvate Kinase from Rabbit Muscle
Shop All MP Biomedicals Products
Click to view available options
Quantity:
100,000 U
50,000 U
Unit Size:
100ku
50ku
Specifications
Specifications
| Concentration | ≥200 U/mg |
| Color | Clear |
| pH | 7.5 |
| For Use With (Application) | Used as a coupling Enzyme in conjunction with Lactic Dehydrogenase in quantifying ADP and the activity of Enzymes that catalyze the formation of ADP |
| Content And Storage | -20°C |
| Source | Rabbit muscle |
| Quantity | 100,000 U |
| Product Type | Pyruvate Kinase |
| Molecular Weight (g/mol) | 237000 |
| Inhibitors | Activity is inhibited by Ca2+, Fluorophosphate inhibits the Enzyme reaction competitively with respect to PEP , ATP inhibits the reaction by removal of Mg2+ from the Substrate MgADP- |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction