Learn More
MP Biomedicals™ Proteinase K from Tritirachium album
Shop All MP Biomedicals ProductsDescription
Proteinase K is used in digestion of proteins from brain tissue samples for prions in Transmissible Spongiform Encephalopathies (TSE) research. Useful for the proteolytic inactivation of nucleases during the isolation of DNA and RNA, as well as in removal of endotoxins bound to cationic proteins such as lysozyme and ribonuclease A and in determination of enzyme localization on membranes.
- Grade: Molecular biology reagent
- Specific Activity: >30 mAnson u/mg
- Presentation: White powder
- Isoelectric Point: 8.9 (Lit.)
- Extinction Coefficient: E1%= 14.2 (280 nm,10 mM NaCl and 5 mM CaCl2, pH 8.0) (Lit.)
- pH: 7.5 -12.00 (denatured hemoglobin as substrate) (Lit.)
- Soluble in water
One Anson unit is the amount of enzyme which will liberate 1 °mole of folin-positive amino acids per minnute at pH 7.5 and 35°C using hemoglobin as substrat
Isolation of DNA and RNA
Specifications
Specifications
| Color | White |
| Grade | Molecular Biology |
| pH | 7.5 to 12 |
| For Use With (Application) | Useful for the proteolytic inactivation of Nucleases during the isolation of DNA and RNA |
| Content And Storage | 4°C |
| Source | Tritirachium album |
| Quantity | 25 mg |
| Product Type | Proteinase K |
| Molecular Weight (g/mol) | 30000 |
| Inhibitors | Diisopropylfluorophosphate or Phenylmethane Sulfonyl Fluoride, AEBSF and Trypsin |
| Show More |
Safety and Handling
- Proteinase K, Molecular Biology Reagent
- Danger
- Serious eye damage/eye irritation Category 2
- Respiratory sensitiser Category 1
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
- H335-May cause respiratory irritation.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P284-¢In case of inadequate ventilation! wear respiratory protection.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P321-Specific treatment (see on this label).
- P405-Store locked up.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
- MIXTURE LIST-Contains : Proteinase K
Your input is important to us. Please complete this form to provide feedback related to the content on this product.