missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MP Biomedicals™ Proteinase K
Shop All MP Biomedicals Products

Click to view available options
Quantity:
1 mg
25 mg
100 mg
250 mg
500 mg
Unit Size:
100mg
1g
250mg
25mg
500mg
Description
Proteinase K, [39450-01-6]
Proteolytic Activity: >30mAnson Units/mg
Unit Definition: One Anson unit is the amount of enzyme which liberates 1 μmol of Folin-positive amino acids per minute at pH 7.5 and 35°C, using hemoglobin as substrate
- From Tritirachium album
- Chromatographically purified
- Exhibits proteolytic activity on proteins, peptides, glycoproteins, amides and esters.
- Active with nitroanilides of amino acids with protected amino groups, excluding arginine.
- Useful in the isolation of DNA and RNA, in the analysis of membrane structures and protein structure.
Specifications
Specifications
| For Use With (Application) | Protein Structure Analysis |
| Molecular Weight (g/mol) | ∽30 |
| Name | Proteinase K |
| pH Range | 7.5 to 12 |
| Purification Method | Chromatography |
| Quantity | 1 mg |
| Source | Tritirachium album Limber |
| Storage Requirements | 4°C |
| Form | Lyophilized |
Safety and Handling
Product Identifier
- Proteinase K
- Danger
- Respiratory sensitiser Category 1
- Skin corrosion/irritation Category 2
- Serious eye damage/eye irritation Category 2
- Specific target organ toxicity Category 3
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
- H335-May cause respiratory irritation.
- P284-¢In case of inadequate ventilation! wear respiratory protection.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P321-Specific treatment (see on this label).
- P405-Store locked up.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction