missing translation for 'onlineSavingsMsg'
Learn More

MP Biomedicals™ Poly-D-Lysine Hydrobromide Attachment Factors

Product Code. 11456632 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
10 mg
25 mg
50 mg
Unit Size:
10mg
25mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11456632 50 mg 50mg
11446632 25 mg 25mg
11436632 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11456632 Supplier MP Biomedicals™ Supplier No. 0215017550
  • ONLINE EXCLUSIVE PRICE
    £224.91
    £193.20 / 50mg
    Save £31.71
    14% Off
 Restricted Item
In Stock
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Facilitate attachment, adherence, and spreading in the culture of adherent cells

Nonsterile.

Specifications

Content And Storage -20°C
Product Type Poly-D-Lysine
Sterility Non-sterile
Quantity 50 mg
Source Synthetic
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.