missing translation for 'onlineSavingsMsg'
Learn More

MP Biomedicals™ Phosphodiesterase I from Crotalus adamanteus

Product Code. 11318559 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
100 U
Unit Size:
100 units
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11318559 100 U 100 units
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11318559 Supplier MP Biomedicals™ Supplier No. 0210097880

Please to purchase this item. Need a web account? Register with us today!

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

Used to breaks phosphodiester bonds. It is a membrane-bound glycoprotein that is used to catalyze the hydrolysis of various nucleotide polyphosphates.

TRUSTED_SUSTAINABILITY

Specifications

Concentration ≥20 μ/mg
pH 9.8 to 10.4.5
For Use With (Application) Used to breaks phosphodiester bonds, catalyze the hydrolysis of various Nucleotide polyphosphates
Content And Storage -20°C
Source Crotalus adamanteus venom
Quantity 100 U
Product Type Phosphodiesterase I
Molecular Weight (g/mol) 115000
Inhibitors Gluctathione, Cysteine and Ascorbic Acid
Form Lyophilized Powder
compliance-icons
Product Identifier
  • Phosphodiesterase I
Signal Word
  • Danger
Hazard Category
  • Respiratory sensitiser Category 1
Hazard Statement
  • H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Precautionary Statement
  • P101-If medical advice is needed, have product container or label at hand.
  • P102-Keep out of reach of children.
  • P103-Read label before use.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P284-[In case of inadequate ventilation] wear respiratory protection.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P342+P311-If experiencing respiratory symptoms: Call a POISON CENTER/doctor/ .
  • P501-Dispose of contents/container to in accordance with all local, regional,national and international regulations.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.