missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MP Biomedicals™ Phosphate Buffered Saline
Shop All MP Biomedicals Products
Click to view available options
Quantity:
10 x 1 L
Unit Size:
10 L
Specifications
Specifications
| Chemical Name or Material | Phosphate Buffered Saline (PBS) |
| pH | 7.4 |
| Quantity | 10 x 1 L |
| Sterility | Sterile |
| Includes | No Calcium, No Magnesium |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction