missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MP Biomedicals™ Peroxidase, 250-330U/mg solid
Shop All MP Biomedicals Products
Click to view available options
Quantity:
10 kU
5 kU
Unit Size:
10ku
5ku
Description
Activity: 250-330U/mg solid. Unit Definition: One unit will form 1mg purpurogallin in 20 seconds from pyrogallol at pH 6.0 and 20°C. Essentially salt-free powder.
Specifications
Specifications
| Concentration | ≥250 U/mg |
| Color | Reddish Brown |
| pH | 7 |
| For Use With (Application) | ELISA, immunohistochemistry, western blotting |
| Content And Storage | 0°C |
| Source | Horseradish |
| Quantity | 5 kU |
| Product Type | Peroxidase |
| Molecular Weight (g/mol) | ≈44000 |
| Inhibitors | HRP is reversibly inhibited by Cyanide and Sulfide at a concentration of 10-5M, p-Aminobenzoic Acid, Azide, Cyclopropanone, L-cystine, Dichromate, Ethylenethiourea, Hydroxylamine, Sulfite and Vanadate and divalent Metals Cd, Co, Cu, Fe, Mn, Ni, Pb |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction