missing translation for 'onlineSavingsMsg'
Learn More

MP Biomedicals™ Peroxidase, 250-330U/mg solid

Product Code. 17158548 Shop All MP Biomedicals Products
Click to view available options
Quantity:
10 kU
5 kU
Unit Size:
10ku
5ku
This item is not returnable. View return policy

Product Code. 17158548

Brand: MP Biomedicals™ 0219537305

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

From horseradish

Activity: 250-330U/mg solid. Unit Definition: One unit will form 1mg purpurogallin in 20 seconds from pyrogallol at pH 6.0 and 20°C. Essentially salt-free powder.

TRUSTED_SUSTAINABILITY

Specifications

Concentration ≥250 U/mg
Color Reddish Brown
pH 7
For Use With (Application) ELISA, immunohistochemistry, western blotting
Content And Storage 0°C
Source Horseradish
Quantity 5 kU
Product Type Peroxidase
Molecular Weight (g/mol) ≈44000
Inhibitors HRP is reversibly inhibited by Cyanide and Sulfide at a concentration of 10-5M, p-Aminobenzoic Acid, Azide, Cyclopropanone, L-cystine, Dichromate, Ethylenethiourea, Hydroxylamine, Sulfite and Vanadate and divalent Metals Cd, Co, Cu, Fe, Mn, Ni, Pb
Form Crystalline Powder
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.