missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MP Biomedicals™ Pepstatin A
Shop All MP Biomedicals Products
Click to view available options
Quantity:
5 mg
10 mg
25 mg
250 mg
Unit Size:
10mg
250mg
25mg
5mg
Description
Pepstatin A, [26305-03-3]; Isoval-Val-Val-Sta-Ala-Sta
C34H63N5O9, F.W. 685.9; Melting Point: 235-237°C
Specifications
Specifications
| For Use With (Application) | Enzyme Assay, Inhibition Assay, Molecular Biology, Protein Purification, Sample Preparation |
| Molecular Weight (g/mol) | 685.91 |
| Name | Pepstatin A |
| Quantity | 250 mg |
| Storage Requirements | 0°C |
| Form | Powder |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction