missing translation for 'onlineSavingsMsg'
Learn More

Penicillin G Potassium Salt, MP Biomedicals™

Product Code. 17690927 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
1 mU
100 μm
25 mU
Unit Size:
1 micro unit
100 micro units
25 micro units
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17690927 100 μm 100 micro units
17670927 1 mU 1 micro unit
17680927 25 mU 25 micro units
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 17690927 Supplier MP Biomedicals Supplier No. 0219453680

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Penicillin G Potassium Salt Cell culture reagent, White Powder

  • It is used as a cell culture additive as an antibiotics.
  • Penicillin G potassium Salt is a salt of Penicillin G.
  • Penicillin G is a hydrophobic, β-lactam antibiotic produced by Penicillium spp.
  • Penicillin G is also known as benzylpenicillin and contains a 6’ phenylacetyl side chain.
  • In acid aqueous solutions, Penicillin is reported to decompose into penillic acid and penilloic acid.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

MDL Number MFCD00036193
Synonym Benzylpenicillin Potassium Salt

Specifications

Physical Form Powder
pH 5 to 7.5 (3% aq. soln.)
Melting Point 214°C to 217°C (Literature)
CAS 113-98-4
Potency 1500 to 1700 μ/mg
Sterility Non-sterile
Color White
Quantity 100 μm
Molecular Formula C16H17N2O4SK
Validated Application Cell Culture
Beilstein 3832841EC
Solubility Information Soluble in water(100mg/mL).
Molecular Weight (g/mol) 372.48
Grade Cell Culture
Assay 0.998
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Penicillin G Potassium Salt, Cell Culture Reagent
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Respiratory sensitiser Category 1
  • Skin sensitiser Category 1
Hazard Statement
  • H302-Harmful if swallowed.
  • H317-May cause an allergic skin reaction.
  • H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P284-[In case of inadequate ventilation] wear respiratory protection.
  • P321-Specific treatment (see on this label).
  • P342+P311-If experiencing respiratory symptoms: Call a POISON CENTER/doctor/ .
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contain: Penicillin G potassium salt
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.