missing translation for 'onlineSavingsMsg'
Learn More

MP Biomedicals™ Papain from Papaya Latex

Product Code. 11344139 Shop All MP Biomedicals Products
Click to view available options
Quantity:
25 mg
500 mg
Unit Size:
25mg
500mg
This item is not returnable. View return policy

Product Code. 11344139

Brand: MP Biomedicals™ 0210092125

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Sulfhydryl protease from Carica papaya latex

Papain cleaves peptide bonds of basic amino acids, leucine, or glycine. It is also hydrolyzes esters and amides.

  • CAS Number: 9001-73-4
  • EC Number: 232-627-2
  • MDL Number: MFCD00131791
  • Format: Milky white solution
  • Specific Activity: ≤10 units/mg Protein
  • Extinction coefficient: 25.0
  • Isoelectric point (pI): pH 9.6
  • Optimum pH: 6.0 to 7.0

Used to produce Fab fragments of antibodies, cell dissociation, morphological isolation of intact cortical neurons from postnatal rats. Limited papaindigestion is used for structural studies of enzymes and other proteins. In addition, papain is used in red cell serology to modify red cell surface to enhance or destroy reactivity of many red cell antigens. Useful for platelet serology studies and has also been used in enzymatic synthesis of amino acids, peptides, and other molecules.

TRUSTED_SUSTAINABILITY

Specifications

Concentration 2X
Color Milky White
pH 6 to 7
For Use With (Application) Produce Fab fragments of Antibodies, cell dissociation, morphological isolation of intact cortical neurons from Postnatal Rats
Content And Storage 4°C
Source Papaya Latex
Quantity 25 mg
Product Type Papain
Molecular Weight (g/mol) 23000
Inhibitors Sulfhydryl groups including heavy Metals, Carbonyl reagents, Aldehydes, Benzoylamidoacetonitrile, AEBSF, antipain, cystatin, E-64, leupeptin, PMSF, TLCK and TPCK
Form Solution
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Papain
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Respiratory sensitiser Category 1
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P101-If medical advice is needed, have product container or label at hand.
  • P102-Keep out of reach of children.
  • P103-Read label before use.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P284-¢In case of inadequate ventilation! wear respiratory protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P321-Specific treatment (see on this label).
  • P405-Store locked up.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contains : Papain
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.