missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
Sulfhydryl protease from Carica papaya latex. (A second protease, chymopapain, and a lysozyme have also been isolated from this same source.)
- Format: Powder
- Grade: Technical
- Molecular weight: 23,000
- Specific Activity:>1750 USP Units/mg
Specifications
Specifications
| Color | Off-White |
| pH | 6 to 7 |
| For Use With (Application) | Enzymatic synthesis of Amino Acids, Peptides and other molecules |
| Content And Storage | 4°C |
| Source | Carica papaya latex |
| Quantity | 100 g |
| Product Type | Papain |
| Molecular Weight (g/mol) | 23000 |
| Inhibitors | AEBSF, Antipain, Cystatin, E-64, leupeptin, PMSF, TLCK and TPCK |
| Form | Powder |
Safety and Handling
Product Identifier
- Papain, Technical Powder
- Danger
- Serious eye damage/eye irritation Category 2
- Respiratory sensitiser Category 1
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
- H335-May cause respiratory irritation.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P284-¢In case of inadequate ventilation! wear respiratory protection.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P321-Specific treatment (see on this label).
- P405-Store locked up.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
- MIXTURE LIST-Contain: papain
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?